3-Fluor-5-(trifluormethyl)phenylboronsäure 99%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Fluoro-5-(trifluoromethyl)benzeneboronic acid, (3-Fluoro-5-(trifluoromethyl)phenyl)boronic acid, 3-(Trifluoromethyl)-5-fluorophenylboronic acid

PC10381-1G PC10381-5G
APOSPC10381-1GEA 27.7 EUR
APOSPC10381-1G APOSPC10381-5G
3-Fluor-5-(trifluormethyl)phenylboronsäure 99%
3-Fluor-5-(trifluormethyl)phenylboronsäure
Formel: C₇H₅BF₄O₂
Molecular Weight: 207.92 g/mol
Schmelzpunkt: 178…180 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06657735
CAS-Nummer: 159020-59-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR