2-Brom-2',3'-difluoracetophenon 98%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 2,3-Difluorophenacyl bromide

PC10408-5G PC10408-10G PC10408-1G
APOSPC10408-5GEA 348 EUR
APOSPC10408-5G APOSPC10408-10G APOSPC10408-1G
2-Brom-2',3'-difluoracetophenon 98%
2-Brom-2',3'-difluoracetophenon
Formel: C₈H₅BrF₂O
Lagertemperatur: Kühlschrank
MDL: MFCD07368687
CAS-Nummer: 886762-77-2
UN: 2920
ADR: 8+3,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR