4-Fluorbenzoylacetonitril

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-(4-Fluorphenyl)-3-oxopropionsäurenitril, 3-(4-Fluorphenyl)-3-ketopropionsäurenitril

PC10409-1G PC10409-5G PC10409-25G
APOSPC10409-1GEA 21.9 EUR
APOSPC10409-1G APOSPC10409-5G APOSPC10409-25G
4-Fluorbenzoylacetonitril
4-Fluorbenzoylacetonitril
Formel: C₉H₆FNO
Molecular Weight: 163.15
Lagertemperatur: Raumtemperatur
MDL: MFCD00662062
CAS-Nummer: 4640-67-9
UN: 3439
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR