2,3-Difluorpyridin 97%

Lieferant: Apollo Scientific
Achtung

PC10418-100G PC10418-25G
APOSPC10418-100GEA 510 EUR
APOSPC10418-100G APOSPC10418-25G
2,3-Difluorpyridin 97%
2,3-Difluoropyridine
Formel: C₅H₃F₂N
Molecular Weight: 115,08 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04114143
CAS-Nummer: 1513-66-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR