2-Fluor-3-nitro-6-picolin 97%
Lieferant: Apollo Scientific
Synonyme:
2-Fluor-6-methyl-3-nitropyridin
PC10420-25G
PC10420-5G
APOSPC10420-25GEA
379
EUR
APOSPC10420-25G
APOSPC10420-5G
2-Fluor-3-nitro-6-picolin 97%
2-Fluor-3-nitro-6-picolin
Formel:
C₆H₅FN₂O₂ Molecular Weight: 156.12 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03095067 CAS-Nummer: 19346-45-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...