4,5-Difluoranthranilsäure 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Amino-4,5-difluorobenzoic acid
PC10504-100G
PC10504-10G
PC10504-25G
APOSPC10504-100GEA
235
EUR
APOSPC10504-100G
APOSPC10504-10G
APOSPC10504-25G
4,5-Difluoranthranilsäure 97%
4,5-Difluoranthranilsäure
Formel:
C₇H₅F₂NO₂ Molecular Weight: 173,12 g/mol Schmelzpunkt: 174…183 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00077509 CAS-Nummer: 83506-93-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...