2-Brom-3,5-difluorpyridin 99%

Lieferant: Apollo Scientific
Achtung

PC10508-1G PC10508-25G PC10508-5G
APOSPC10508-1GEA 26.1 EUR
APOSPC10508-1G APOSPC10508-25G APOSPC10508-5G
2-Brom-3,5-difluorpyridin 99%
2-Bromo-3,5-difluoropyridine
Formel: C₅H₂BrF₂N
Molecular Weight: 193.98 g/mol
Siedepunkt: 36…37 °C/0.6mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD03840197
CAS-Nummer: 660425-16-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR