2-Fluor-5-methoxybenzylalkohol

Lieferant: Apollo Scientific
Achtung

Synonyme: (2-Fluoro-5-methoxyphenyl)methanol

PC1052-1G PC1052-25G PC1052-5G
APOSPC1052-1GEA 34.8 EUR
APOSPC1052-1G APOSPC1052-25G APOSPC1052-5G
2-Fluor-5-methoxybenzylalkohol
2-Fluor-5-methoxybenzylalkohol
Formel: C₈H₉FO₂
Molecular Weight: 156,16 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04116337
CAS-Nummer: 161643-29-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR