3-Fluorophenylglyoxal hydrate 97%
Lieferant: Apollo Scientific
PC10535-25G
PC10535-5G
PC10535-1G
PC10535-10G
APOSPC10535-25GEA
0
EUR
APOSPC10535-25G
APOSPC10535-5G
APOSPC10535-1G
APOSPC10535-10G
3-Fluorophenylglyoxal hydrate 97%
3-Fluorophenylglyoxal hydrate
Formel:
C₈H₇O₃F Lagertemperatur: Raumtemperatur |
MDL:
MFCD06800769 CAS-Nummer: 121247-01-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...