2-Fluorchinolin-3-boronsäure 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC10538-1G
PC10538-250MG
APOSPC10538-1GEA
314
EUR
APOSPC10538-1G
APOSPC10538-250MG
2-Fluorchinolin-3-boronsäure 97%
2-Fluorchinolin-3-boronsäure
Formel:
C₉H₇BFNO₂ Molecular Weight: 190.97 g/mol Schmelzpunkt: 89…94 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD05664131 CAS-Nummer: 745784-10-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...