(4-Aminophenyl){4-[4-(trifluoromethyl)phenyl]piperazino}methanone

Lieferant: Apollo Scientific
Achtung

PC10558-1G PC10558-250MG
APOSPC10558-1GEA 186 EUR
APOSPC10558-1G APOSPC10558-250MG
(4-Aminophenyl){4-[4-(trifluoromethyl)phenyl]piperazino}methanone
(4-Aminophenyl){4-[4-(trifluoromethyl)phenyl]piperazino}methanone
Formel: C₁₈H₁₈F₃N₃O
Molecular Weight: 349,36 g/mol
Schmelzpunkt: 151 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06659566
CAS-Nummer: 885949-76-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR