4'-Benzyloxy-3'-fluoracetophenon
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-(4-(Benzyloxy)-3-fluorophenyl)ethanone
PC10559-1G
PC10559-5G
PC10559-250MG
APOSPC10559-1GEA
140
EUR
APOSPC10559-1G
APOSPC10559-5G
APOSPC10559-250MG
4'-Benzyloxy-3'-fluoracetophenon
4'-Benzyloxy-3'-fluoracetophenon
Formel:
C₁₅H₁₃FO₂ Molecular Weight: 244,261 g/mol Schmelzpunkt: 79…81 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06659550 CAS-Nummer: 81227-99-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...