1-[4-(Trifluoromethoxy)phenyl]-1H-pyrrole-2,5-dione

Lieferant: Apollo Scientific
Achtung

Synonyme: N-[4-(Trifluoromethoxy)phenyl]maleimide, 1-(4-(Trifluoromethoxy)phenyl)-1H-pyrrole-2,5-dione

PC10592-1G PC10592-5G
APOSPC10592-1GEA 85.7 EUR
APOSPC10592-1G APOSPC10592-5G
1-[4-(Trifluoromethoxy)phenyl]-1H-pyrrole-2,5-dione
1-[4-(Trifluoromethoxy)phenyl]-1H-pyrrole-2,5-dione
Formel: C₁₁H₆F₃NO₃
Molecular Weight: 257.17 g/mol
Schmelzpunkt: 76…77 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00106927
CAS-Nummer: 68255-58-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR