4-Cyan-3-fluoranilin 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC10597-1G
PC10597-25G
PC10597-5G
APOSPC10597-1GEA
21.9
EUR
APOSPC10597-1G
APOSPC10597-25G
APOSPC10597-5G
4-Cyan-3-fluoranilin 95%
4-Cyan-3-fluoranilin
Formel:
C₇H₅FN₂ Lagertemperatur: Kühlschrank |
MDL:
MFCD07368709 CAS-Nummer: 53312-80-4 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...