2-Methyl-4-(trifluormethoxy)benzaldehyd 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC1062-1G
PC1062-5G
APOSPC1062-1GEA
45.1
EUR
APOSPC1062-1G
APOSPC1062-5G
2-Methyl-4-(trifluormethoxy)benzaldehyd 98%
2-Methyl-4-(trifluoromethoxy)benzaldehyde
Formel:
C₉H₇F₃O₂ Molecular Weight: 204,15 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07368717 CAS-Nummer: 886763-07-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...