3-Bromo-4-(trifluoromethyl)benzyl alcohol

Lieferant: Apollo Scientific
Achtung

Synonyme: Benzenemethanol, 3-bromo-4-(trifluoromethyl)

PC10670-1G PC10670-5G
APOSPC10670-1GEA 46.4 EUR
APOSPC10670-1G APOSPC10670-5G
3-Bromo-4-(trifluoromethyl)benzyl alcohol
3-Bromo-4-(trifluoromethyl)benzyl alcohol
Formel: C₈H₆BrF₃O
Molecular Weight: 255,04 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13185397
CAS-Nummer: 372120-54-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR