Tris(2,2′-bipyridine)ruthenium(II) hexafluorophosphate 99%
Lieferant: Apollo Scientific
Synonyme: Tris(2,2'-bipyridine)ruthenium bis(hexafluorophosphate), Ruthenium-tris(2,2′-bipyridyl) dihexafluorophosphate, Ru(bpy)₃(PF₆)₂, Tris(2,2'-bipyridine)ruthenium(II) Bis(hexafluorophosphate)
PC10698-1G
APOSPC10698-1GEA 172 EUR
APOSPC10698-1G
Tris(2,2′-bipyridine)ruthenium(II) hexafluorophosphate 99%
Tris(2,2′-bipyridine)ruthenium(II) hexafluorophosphate
Formel: C₃₀H₂₄F₁₂N₆P₂Ru
Molecular Weight: 859,562 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11042502
CAS-Nummer: 60804-74-2