2-Amino-3-chlor-5-(trifluormethyl)pyridin 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC1070G-10G
PC1070G-25G
APOSPC1070G-10GEA
14.7
EUR
APOSPC1070G-10G
APOSPC1070G-25G
2-Amino-3-chlor-5-(trifluormethyl)pyridin 97%
2-Amino-3-chloro-5-(trifluoromethyl)pyridine
Formel:
C₆H₄ClF₃N₂ Molecular Weight: 196,5600 g/mol Schmelzpunkt: 86…96 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00042154 CAS-Nummer: 79456-26-1 EINECS: 401-670-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...