3-Amino-2-(2,4-difluorophenoxy)pyridine 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-(2,4-Difluorophenoxy)pyridin-3-amine
PC1070R-250MG
PC1070R-1G
PC1070R-5G
APOSPC1070R-250MGEA
36.4
EUR
APOSPC1070R-250MG
APOSPC1070R-1G
APOSPC1070R-5G
3-Amino-2-(2,4-difluorophenoxy)pyridine 98%
3-Amino-2-(2,4-difluorophenoxy)pyridine
Formel:
C₁₁H₈F₂N₂O Molecular Weight: 222,2 g/mol Schmelzpunkt: 95…7 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00067802 CAS-Nummer: 175135-63-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...