4-Fluoranthranilsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Amino-4-fluorobenzoic acid
PC1071C-100G
PC1071C-250G
PC1071C-25G
PC1071C-500G
APOSPC1071C-100GEA
89.9
EUR
APOSPC1071C-100G
APOSPC1071C-250G
APOSPC1071C-25G
APOSPC1071C-500G
4-Fluoranthranilsäure 98%
4-Fluoranthranilsäure
Formel:
C₇H₆FNO₂ Molecular Weight: 155,13 g/mol Schmelzpunkt: 192…196 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00075553 CAS-Nummer: 446-32-2 EINECS: 207-163-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...