5-Fluoranthranilsäure 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Amino-5-fluorobenzoic acid

PC1071F-100G
APOSPC1071F-100GEA 97.3 EUR
APOSPC1071F-100G
5-Fluoranthranilsäure 97%
5-Fluoranthranilsäure
Formel: C₇H₆FNO₂
Molecular Weight: 155,13 g/mol
Schmelzpunkt: 181…183 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00055566
CAS-Nummer: 446-08-2
EINECS: 207-159-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR