Sodium trifluoromethanesulphinate 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Trifluoromethanesulphinic acid Sodium Salt, Langlois Reagent
PC10732-1G
PC10732-25G
PC10732-5G
PC10732-100G
APOSPC10732-1GEA
20.5
EUR
APOSPC10732-1G
APOSPC10732-25G
APOSPC10732-5G
APOSPC10732-100G
Sodium trifluoromethanesulphinate 98%
Sodium trifluoromethanesulphinate
Formel:
CHF₃O₂S Molecular Weight: 156,05 g/mol Schmelzpunkt: 350 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03092989 CAS-Nummer: 2926-29-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...