3-Amino-2-(4-fluorphenoxy)pyridin 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-(4-Fluorophenoxy)pyridin-3-amine
PC1076O-1G
PC1076O-250MG
PC1076O-500MG
PC1076O-2.5G
APOSPC1076O-1GEA
163
EUR
APOSPC1076O-1G
APOSPC1076O-250MG
APOSPC1076O-500MG
APOSPC1076O-2.5G
3-Amino-2-(4-fluorphenoxy)pyridin 98%
3-Amino-2-(4-fluorophenoxy)pyridine
Formel:
C₁₁H₉FN₂O Schmelzpunkt: 101…103 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00067803 CAS-Nummer: 175135-64-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...