3-Amino-4-isopropylaminobenzotrifluorid 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC1076Z-1G
PC1076Z-250MG
APOSPC1076Z-1GEA
81.2
EUR
APOSPC1076Z-1G
APOSPC1076Z-250MG
3-Amino-4-isopropylaminobenzotrifluorid 97%
3-Amino-4-isopropylaminobenzotrifluorid
Formel:
C₁₀H₁₃F₃N₂ Schmelzpunkt: 63…65 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00084879 CAS-Nummer: 175277-91-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...