3-Fluoranthranilsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Amino-3-fluorobenzoic acid

PC1089-1G PC1089-5G PC1089-25G
APOSPC1089-1GEA 21.9 EUR
APOSPC1089-1G APOSPC1089-5G APOSPC1089-25G
3-Fluoranthranilsäure
3-Fluoranthranilsäure
Formel: C₇H₆FNO₂
Molecular Weight: 155,13 g/mol
Schmelzpunkt: 185…187 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01569395
CAS-Nummer: 825-22-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR