4-Nitro-3-(trifluormethyl)anilin 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Amino-2-nitro-α,α,α-trifluortoluol, 4-Nitro-α,α,α-trifluor-m-toluidin

PC1094-100G PC1094-25G
APOSPC1094-100GEA 26.1 EUR
APOSPC1094-100G APOSPC1094-25G
4-Nitro-3-(trifluormethyl)anilin 98%
4-Nitro-3-(trifluormethyl)anilin
Formel: C₇H₅F₃N₂O₂
Molecular Weight: 206,12 g/mol
Schmelzpunkt: 125…129 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00014717
CAS-Nummer: 393-11-3
EINECS: 206-884-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR