2-Fluor-5-(trifluormethoxy)benzonitril 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC1097-5G
PC1097-10G
PC1097-1G
APOSPC1097-5GEA
89.9
EUR
APOSPC1097-5G
APOSPC1097-10G
APOSPC1097-1G
2-Fluor-5-(trifluormethoxy)benzonitril 97%
2-Fluor-5-(trifluormethoxy)benzonitril
Formel:
C₈H₃F₄NO Molecular Weight: 205,11 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04115894 CAS-Nummer: 886498-08-4 UN: 3276 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...