2-Amino-4,4,4-trifluorbuttersäure 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(±)-2-Amino-4,4,4-trifluorbuttersäure
PC1100A-1G
PC1100A-250MG
PC1100A-5G
APOSPC1100A-1GEA
313
EUR
APOSPC1100A-1G
APOSPC1100A-250MG
APOSPC1100A-5G
2-Amino-4,4,4-trifluorbuttersäure 97%
2-Amino-4,4,4-trifluorbuttersäure
Formel:
C₄H₆F₃NO₂ Molecular Weight: 157,09 g/mol Schmelzpunkt: 260…265 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00041417 CAS-Nummer: 15959-93-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...