2-Amino-6-(trifluoromethoxy)benzothiazole 97%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
6-(Trifluoromethoxy)benzo[d]thiazol-2-amine, Riluzole
PC1100C-1G
APOSPC1100C-1GEA
21.9
EUR
APOSPC1100C-1G
2-Amino-6-(trifluoromethoxy)benzothiazole 97%
2-Amino-6-(trifluoromethoxy)benzothiazole
Formel:
C₈H₅F₃N₂OS Molecular Weight: 234,20 g/mol Schmelzpunkt: 114…118 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00210213 CAS-Nummer: 1744-22-5 EINECS: 605-724-6 UN: 2811 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...