7-Amino-4-(trifluormethyl)cumarin 99%

Lieferant: Apollo Scientific
Achtung

Synonyme: 7-Amino-4-(trifluoromethyl)coumarin, 7-Amino-4-trifluoromethylcoumarin, 7-Amino-4-(trifluoromethyl)-2H-chromen-2-one, Coumarin 151

PC1100CF-10G PC1100CF-1G PC1100CF-5G
APOSPC1100CF-10GEA 142 EUR
APOSPC1100CF-10G APOSPC1100CF-1G APOSPC1100CF-5G
7-Amino-4-(trifluormethyl)cumarin 99%
7-Amino-4-(trifluormethyl)cumarin

Useful intermediate for the synthesis of substrates used in fluorometric assay of proteolytic enzymes; Anal. Biochem.,70,258 (1976)

Formel: C₁₀H₆F₃NO₂
Molecular Weight: 229,16 g/mol
Schmelzpunkt: 221…223 °C
Dichte: 1,59 g/cm³ (25 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00006858
CAS-Nummer: 53518-15-3
EINECS: 258-599-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR