2-Amino-5-(trifluormethyl)-1,3,4-thiadiazol 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Amino-5-trifluoromethyl-1,3,4-thiadiazole

PC1100J-1G PC1100J-25G PC1100J-5G
APOSPC1100J-1GEA 21.9 EUR
APOSPC1100J-1G APOSPC1100J-25G APOSPC1100J-5G
2-Amino-5-(trifluormethyl)-1,3,4-thiadiazol 98%
2-Amino-5-(trifluoromethyl)-1,3,4-thiadiazole
Formel: C₃H₂F₃N₃S
Molecular Weight: 169,13 g/mol
Schmelzpunkt: 225…227 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00003109
CAS-Nummer: 10444-89-0
EINECS: 233-930-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR