(2-Chlor-4-fluorphenyl)methansulfonsäurechlorid

Lieferant: Apollo Scientific

PC110117-1G PC110117-250MG
APOSPC110117-1GEA 174 EUR
APOSPC110117-1G APOSPC110117-250MG
(2-Chlor-4-fluorphenyl)methansulfonsäurechlorid
(2-Chloro-4-fluorophenyl)methanesulphonyl chloride
Formel: C₇H₅Cl₂FO₂S
Lagertemperatur: Raumtemperatur
CAS-Nummer: 541528-45-4
UN: 3261
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR