4-Fluor-2-methoxybenzonitril 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Fluor-2-methoxybenzoesäurenitril

PC1109-25G PC1109-5G
APOSPC1109-25GEA 212 EUR
APOSPC1109-25G APOSPC1109-5G
4-Fluor-2-methoxybenzonitril 97%
4-Fluor-2-methoxybenzonitril
Formel: C₈H₆FNO
Molecular Weight: 151.14 g/mol
Schmelzpunkt: 75…77 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04116335
CAS-Nummer: 191014-55-8
UN: 3439
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR