2-Brom-4-(trifluormethyl)benzonitril

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Bromo-4-(trifluoromethyl)benzonitile

PC11108-5G PC11108-25G PC11108-100G
APOSPC11108-5GEA 23.2 EUR
APOSPC11108-5G APOSPC11108-25G APOSPC11108-100G
2-Brom-4-(trifluormethyl)benzonitril
2-Brom-4-(trifluormethyl)benzonitril
Formel: C₈H₃BrF₃N
Molecular Weight: 250,0200 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06659467
CAS-Nummer: 35764-15-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR