4-Fluor-2-methoxyphenylboronsäure 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Fluoro-2-methoxybenzeneboronic acid

PC11112-25G
APOSPC11112-25GEA 155 EUR
APOSPC11112-25G
4-Fluor-2-methoxyphenylboronsäure 98%
4-Fluor-2-methoxyphenylboronsäure
Formel: C₇H₈BFO₃
Molecular Weight: 169,95 g/mol
Schmelzpunkt: 131…136 °C
Lagertemperatur: Kühlschrank
MDL: MFCD03788423
CAS-Nummer: 179899-07-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR