5-Fluor-2-methoxyphenylboronsäure
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Fluoro-2-methoxybenzeneboronic acid
PC11113-100G
PC11113-25G
PC11113-5G
APOSPC11113-100GEA
151
EUR
APOSPC11113-100G
APOSPC11113-25G
APOSPC11113-5G
5-Fluor-2-methoxyphenylboronsäure
5-Fluor-2-methoxyphenylboronsäure
Formel:
C₇H₈BFO₃ Molecular Weight: 169,95 g/mol Schmelzpunkt: 152…158 °C Flammpunkt: >110 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD01863526 CAS-Nummer: 179897-94-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...