3-Fluor-4-methylphenylboronsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Fluoro-4-methylbenzeneboronic acid

PC11114-1G PC11114-25G PC11114-5G
APOSPC11114-1GEA 14.7 EUR
APOSPC11114-1G APOSPC11114-25G APOSPC11114-5G
3-Fluor-4-methylphenylboronsäure
3-Fluor-4-methylphenylboronsäure
Formel: C₇H₈BFO₂
Molecular Weight: 153,95 g/mol
Schmelzpunkt: 234…237 °C
Lagertemperatur: Kühlschrank
MDL: MFCD02683115
CAS-Nummer: 168267-99-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR