4-Fluor-1-naphthalinboronsäure
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Fluoronaphtalene-1-boronic acid, 4-Fluoronaphthalene-1-boronic acid
PC11115-1G
PC11115-250MG
PC11115-5G
APOSPC11115-1GEA
74.1
EUR
APOSPC11115-1G
APOSPC11115-250MG
APOSPC11115-5G
4-Fluor-1-naphthalinboronsäure
4-Fluoro-1-naphthaleneboronic acid
Formel:
C₁₀H₇BBrFO₂ Molecular Weight: 189,98 g/mol Schmelzpunkt: 240…242 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04039221 CAS-Nummer: 182344-25-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...