4'-Fluoro-biphenyl-3-ylamine

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Amino-4'-fluorobiphenyl hydrochloride

PC11127-1G PC11127-5G
APOSPC11127-1GEA 230 EUR
APOSPC11127-1G APOSPC11127-5G
4'-Fluoro-biphenyl-3-ylamine
4'-Fluoro-biphenyl-3-ylamine
Formel: C₁₂H₁₀FN
Molecular Weight: 187.22 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03424674
CAS-Nummer: 10540-45-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR