3'-Fluorobiphenyl-4-carbaldehyde

Lieferant: Apollo Scientific
Achtung

Synonyme: 3'-Fluoro-[1,1'-biphenyl]-4-carbaldehyde

PC11134-1G PC11134-5G PC11134-100MG
APOSPC11134-1GEA 132 EUR
APOSPC11134-1G APOSPC11134-5G APOSPC11134-100MG
3'-Fluorobiphenyl-4-carbaldehyde
3'-Fluorobiphenyl-4-carbaldehyde
Formel: C₁₃H₉FO
Molecular Weight: 200,21 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04039225
CAS-Nummer: 400750-63-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR