2-(4-Fluorphenyl)benzaldehyd
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4'-Fluorobiphenyl-2-carboxaldehyde, 4'-Fluoro-[1,1'-biphenyl]-2-carbaldehyde
PC11135-1G
PC11135-250MG
APOSPC11135-1GEA
495
EUR
APOSPC11135-1G
APOSPC11135-250MG
2-(4-Fluorphenyl)benzaldehyd
2-(4-Fluorophenyl)benzaldehyde
Formel:
C₁₃H₉FO Molecular Weight: 200,21 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01631854 CAS-Nummer: 192863-46-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...