3'-Fluorbiphenyl-2-carbonsäure

Lieferant: Apollo Scientific
Achtung

PC11139-1G PC11139-250MG
APOSPC11139-1GEA 128 EUR
APOSPC11139-1G APOSPC11139-250MG
3'-Fluorbiphenyl-2-carbonsäure
3'-Fluorbiphenyl-2-carbonsäure
Formel: C₁₃H₉FO₂
Molecular Weight: 216.21
Schmelzpunkt: 96…98 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03426488
CAS-Nummer: 2094-03-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR