4'-Fluorbiphenyl-4-carbonsäure 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4'-Fluoro-[1,1'-biphenyl]-4-carboxylic acid, 4-(4-Fluorophenyl)benzoic acid
PC11144-1G
PC11144-5G
APOSPC11144-1GEA
127
EUR
APOSPC11144-1G
APOSPC11144-5G
4'-Fluorbiphenyl-4-carbonsäure 95%
4'-Fluorbiphenyl-4-carbonsäure
Formel:
C₁₃H₉FO₂ Molecular Weight: 216,21 g/mol Schmelzpunkt: 235…240 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01631909 CAS-Nummer: 5731-10-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...