3'-(Trifluoromethyl)biphenyl-4-carboxaldehyde
Lieferant: Apollo Scientific
|
Gefahr
|
PC11176-1G
PC11176-5G
APOSPC11176-1GEA
132
EUR
APOSPC11176-1G
APOSPC11176-5G
3'-(Trifluoromethyl)biphenyl-4-carboxaldehyde
3'-(Trifluoromethyl)biphenyl-4-carboxaldehyde
Formel:
C₁₄H₉F₃O Molecular Weight: 250,22 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03424645 CAS-Nummer: 100036-64-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...