2,3-Difluor-6-methoxybenzoesäure 98%

Lieferant: Apollo Scientific
Achtung

PC1118-1G PC1118-25G PC1118-5G
APOSPC1118-1GEA 52.2 EUR
APOSPC1118-1G APOSPC1118-25G APOSPC1118-5G
2,3-Difluor-6-methoxybenzoesäure 98%
2,3-Difluoro-6-methoxybenzoic acid
Formel: C₈H₆F₂O₃
Molecular Weight: 188.13 g/mol
Schmelzpunkt: 126…129 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04116018
CAS-Nummer: 773873-26-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR