1-[2-Fluoro-4-(methylsulphonyl)phenyl]piperazine 95+%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-(2-Fluoro-4-(methylsulphonyl)phenyl)piperazine
PC11194-1G
PC11194-5G
PC11194-10G
PC11194-250MG
APOSPC11194-1GEA
132
EUR
APOSPC11194-1G
APOSPC11194-5G
APOSPC11194-10G
APOSPC11194-250MG
1-[2-Fluoro-4-(methylsulphonyl)phenyl]piperazine 95+%
1-[2-Fluoro-4-(methylsulphonyl)phenyl]piperazine
Formel:
C₁₁H₁₅FN₂O₂S Molecular Weight: 258.32 g/mol Schmelzpunkt: 115…122 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03648602 CAS-Nummer: 845616-10-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...