4-Fluoro-2-methyl-1-(methylsulphonyl)benzene 95+%
Lieferant: Apollo Scientific
|
Achtung
|
PC11207-1G
PC11207-5G
PC11207-25G
APOSPC11207-1GEA
208
EUR
APOSPC11207-1G
APOSPC11207-5G
APOSPC11207-25G
4-Fluoro-2-methyl-1-(methylsulphonyl)benzene 95+%
4-Fluoro-2-methyl-1-(methylsulphonyl)benzene
Formel:
C₈H₉FO₂S Schmelzpunkt: 104…105 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04037935 CAS-Nummer: 828270-62-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...