1-Fluoro-5-methyl-4-(methylsulphonyl)-2-nitrobenzene 95+%

Lieferant: Apollo Scientific

PC11208-1G PC11208-5G PC11208-25G
APOSPC11208-1GEA 229 EUR
APOSPC11208-1G APOSPC11208-5G APOSPC11208-25G
1-Fluoro-5-methyl-4-(methylsulphonyl)-2-nitrobenzene 95+%
1-Fluoro-5-methyl-4-(methylsulphonyl)-2-nitrobenzene
Formel: C₈H₈FNO₄S
Schmelzpunkt: 150…151 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04037941
CAS-Nummer: 849035-74-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR