4,5-Difluor-2-methoxybenzaldehyd
Lieferant: Apollo Scientific
|
Achtung
|
PC1122-1G
PC1122-5G
PC1122-250MG
APOSPC1122-1GEA
30.6
EUR
APOSPC1122-1G
APOSPC1122-5G
APOSPC1122-250MG
4,5-Difluor-2-methoxybenzaldehyd
4,5-Difluor-2-methoxybenzaldehyd
Formel:
C₈H₆F₂O₂ Schmelzpunkt: 74 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06246884 CAS-Nummer: 145742-34-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...