Kalium(3-chlor-4-fluorphenyl)trifluoroborat
Lieferant: Apollo Scientific
Synonyme:
(3-Chlor-4-fluorphenyl)trifluorborsäure Kaliumsalz
PC11231-1G
PC11231-5G
PC11231-25G
APOSPC11231-1GEA
79.9
EUR
APOSPC11231-1G
APOSPC11231-5G
APOSPC11231-25G
Kalium(3-chlor-4-fluorphenyl)trifluoroborat
Kalium(3-chlor-4-fluorphenyl)trifluoroborat
Formel:
C₆H₃BClF₄K Molecular Weight: 236,44 g/mol Schmelzpunkt: 234…244 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04115752 CAS-Nummer: 850623-59-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...